8270 Matrix Spike Mix (76 components)
: Restek
The analysis of semivolatile organic pollutants in solid waste, soil, water, and air matrices using gas chromatography - mass spectrometry (GC - MS).
1,2-diphenylhydrazine decomposes to azobenzene (mix component) in the injector. N-nitrosodiphenylamine decomposes to diphenylamine (mix component) in the injector.
: EPA standards.
Frequently Bought Together