Order Entry
Switzerland
ContactUsLinkComponent
Ammonium thiocyanate 0.02 mol/l (0.02 N) in aqueous solution volumetric solution
Ammonium thiocyanate 0.02 mol/l (0.02 N) in aqueous solution volumetric solution
Catalog # 302634.1000
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ammonium thiocyanate 0.02 mol/l (0.02 N) in aqueous solution volumetric solution
Catalog # 302634.1000
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :76.12 g/mol
Storage Temperature:Ambient
EINECS:217-175-6
CAS Number:1762-95-4
Formula:NH₄SCN
MDL Number:MFCD00011428