Order Entry
Switzerland
ContactUsLinkComponent
Sodium chloride 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Sodium chloride 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 85811.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium chloride 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 85811.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :58.44 g/mol
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477