Order Entry
Switzerland
ContactUsLinkComponent
Nitric acid 2 mol/l (2 N), AVS TITRINORM volumetric solution
Corrosive
Nitric acid 2 mol/l (2 N), AVS TITRINORM volumetric solution
Catalog # 310568.1000
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Nitric acid 2 mol/l (2 N), AVS TITRINORM volumetric solution
Catalog # 310568.1000
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :63.01 g/mol
Storage Temperature:Ambient
UN:2031
ADR:8,II
 :GHS05
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349