Sodium nitrite 0.1 N, AVS TITRINORM volumetric solution
Catalog # 89295.320
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
About this item
:
:69 g/mol
CAS Number:7632-00-0
Formula:NaNO₂
MDL Number:MFCD00011118