ContactUsLinkComponent
Disinfectant detergent, SURE® Ice & Shake
Disinfectant detergent, SURE® Ice & Shake
  DIVY100913010
 :  Diversey
   Safer Human and Environmental Health
   Sustainable Materials
undefined
Disinfectant detergent, SURE® Ice & Shake
  DIVY100913010
 :  Diversey
   Safer Human and Environmental Health
   Sustainable Materials
  • Description:
    SURE® Ice & Shake, detergent disinfectant
  • Seq:
    02
  • Environmentally Preferable:
    Safer Human and Environmental Health,
    Sustainable Materials
  • Environmental certification:
    OECD 301B
    Cradle to creadle certification at gold level
  • Packed:
    1 litre bottle
  • Pk:
    6 Bottle
SURE® Ice and shake is a concentrated detergent disinfectant suitable for the cleaning, disinfection and milkstone removal of shake, ice machines and other beverage equipment found in food service premises.
  • Saves time; cleaning, disinfection and milkstone removal is done with one single product in one application
  • Very effective against a wide range of microorganisms; bactericidal and yeasticidal activities
  • Contains plant based actives from renewable sources decomposed by natural processes
  • No environmental hazard classifications, no safety warnings for product in use for user
  • No fragrance nor dye, natural odour present due to plant based ingredients
SURE® Ice & Shake is part of a unique range of plant-based, cleaning products that deliver the professional cleaning results you need - while being safe for people and kind to the environment. SURE® natural cleaning products provide safety and sustainability without compromising on performance. Bottles/cans suitable for recycling.
 : OECD 301B. Cradle to cradle certification at gold level.