



n-Hexane, PESTINORM® Supra Trace for MOSH/MOAH analysis
Catalog # 85957.320
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Appearance:Clear colorless liquid
- Identity (IR):Complies
- Color (APHA, Pt-Co):max. 10
- Assay (GC, on anhydrous basis):min. 99.0 %
- Hexane isomers + methylcyclopentane:min. 99.8 %
- MOSH Test by GC/FID:max. 5 ng/ml
- MOAH Test by LC/UV:max. 5 ng/ml
- GC/ECD 1,2,4-TCB to Deca-PCB (as Lindane):max. 3 pg/ml
- Water (KF):max. 0.02 % w/w
- Residue after evaporation:max. 0.0003 % w/w
Specification Test Results
About this item
:
:86.18 g/mol
Merck Index:13,04712
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–94,3 °C
Density:0.659 g/cm³ (20 °C)
Flash Pt:–22 °C
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
:Filtered through a 0.2 µm filter, packaged under inert gas.
:GHS02
:GHS08
:GHS06
:GHS09
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520