Order Entry
Switzerland
ContactUsLinkComponent
Potassium chlorate ≥99.0% (by KMnO₄ titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Oxidizing
Irritant
Environment
Potassium chlorate ≥99.0% (by KMnO₄ titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 3024-07
Supplier:  Avantor
CAS Number:  
undefined
Oxidizing
Irritant
Environment
Potassium chlorate ≥99.0% (by KMnO₄ titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 3024-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE pail, screw top
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • Assay (KClO₃) (by KMnO₄ titrn):
    ≥99.0 %
  • Insoluble Matter:
    ≤0.005 %
  • pH of 5% Solution at 25°C, For Information Only:
    For Information Only
  • Bromate (BrO₃):
    ≤0.01 %
  • Calcium (Ca):
    ≤0.002 %
  • Chloride (Cl):
    ≤0.001 %
  • ACS - Magnesium (Mg):
    ≤0.002 %
  • Sulfate (SO₄):
    Passes Test
  • Sodium (Na):
    ≤0.010 %
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤2.0 ppm

Specification Test Results

About this item

 :
 :122.55 g/mol
Merck Index:13,07703
Boiling Pt:400 °C (1013 hPa)
Melting Pt:356 °C
Density:2.34 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:223-289-7
UN:1485
ADR:5.1,II
 :GHS03
 :GHS07
 :GHS09
CAS Number:3811-04-9
Formula:KClO₃
MDL Number:MFCD00011361