Hand sanitiser, antibacterial hand wash Free, SURE®
Catalog # DIVY101100797
Supplier: Diversey
Specifications
- Description:SURE® antibacterial hand wash free, 1,3 L
- Seq:4
- Environmentally Preferable:Sustainable Materials,Safer Human and Environmental Health,Sustainable Packaging
- Pk:4 Bottle
Specifications
About this item
Cleans and disinfects hands in one easy step. Based on lactic acid, an organic based ingredient from Natural sources that inhibits the growth of micro-organisms including bacteria, yeasts and some viruses.
- Contains plant based ingredients from renewable sources
- Decomposed by natural processes
- Passes EN test standards: EN 1499, EN 13727 and EN13624 (yeast only), EN 1276, EN 1650 EN14476 (Influenza A/H1N1) similar to the SARS-CoV-2 virus
- Passes the latest BPR test requirements for hand disinfection
- Triclosan- and paraben-free
- No dyes
- Proven to not taint food
- Extensively proven skin mildness (ROAT, TEWL and Patch tests)
- No environmental hazard classifications, and no safety warnings for users
SURE® is a comprehensive offering of plant-based, cleaning products designed to deliver superior professional results while being safe for people and kind to the environment.
With environmentally-friendly ingredients from renewable resources, SURE® products meet the daily requirements for kitchens, facilities, and personal care.
Certifications: According to OECD 301B test. Cradle to cradle certification at gold level.
Ordering information: 1,3 L cartridge, suitable for Diversey IntelliCare Dispenser (manual/hybrid). 800 ml cartridge is suitable for Diversey Soft Care Line dispenser.
Caution: Use biocides safely. Always read the label and product information before use.