Order Entry
Switzerland
ContactUsLinkComponent
 
Analytical standard, Flucloxacillin Natriumsalz, reference material, Honeywell Fluka™
Catalog # HONCR1331-100MG
CAS Number:  
Analytical standard, Flucloxacillin Natriumsalz, reference material, Honeywell Fluka™
Catalog # HONCR1331-100MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    100 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1847-24-1
  • Description:
    Flucloxacillin Natriumsalz
  • Melting point:
    184 °C
  • Formula:
    C₁₉H₁₆ClFN₃NaO₅S
  • Storage temperature:
    2…8 °C
  • Description:
    Flucloxacillin Natriumsalz

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results