Analytical standard, Propachlor-(isopropyl-d₇), reference material, Honeywell Fluka™
HONCR1301-10MG
:
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- Description:Propachlor-(isopropyl-d₇)
- Melting point:67…69 °C
- Formula:C₁₁D₇H₇ClNO
- Storage temperature:Ambient
- Description:Propachlor-(isopropyl-d₇)
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results