Order Entry
Switzerland
ContactUsLinkComponent
Analytical standard, Salbutamol-(tert-butyl-d₉) acetate, reference material, Honeywell Fluka™
Analytical standard, Salbutamol-(tert-butyl-d₉) acetate, reference material, Honeywell Fluka™
Catalog # HONCR1211-10MG
CAS Number:  
Analytical standard, Salbutamol-(tert-butyl-d₉) acetate, reference material, Honeywell Fluka™
Catalog # HONCR1211-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1173021-73-2
  • Description:
    Salbutamol-(tert-butyl-d₉) acetate
  • Melting point:
    158 °C
  • Formula:
    C₁₃D₉H₁₂NO₃ · C₂H₄O₂
  • Storage temperature:
    Ambient
  • Description:
    Salbutamol-(tert-butyl-d₉) acetate

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results