Order Entry
Switzerland
ContactUsLinkComponent
Analytical standard, Imidocarb dihydrochloride monohydrate, reference material, Honeywell Fluka™
Analytical standard, Imidocarb dihydrochloride monohydrate, reference material, Honeywell Fluka™
Catalog # HONCR1206-10MG
CAS Number:  
Analytical standard, Imidocarb dihydrochloride monohydrate, reference material, Honeywell Fluka™
Catalog # HONCR1206-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    5318-76-3
  • Description:
    Imidocarb dihydrochloride monohydrate
  • Melting point:
    300 °C
  • Formula:
    C₁₉H₂₀N₆O · 2HCl · H₂O
  • Storage temperature:
    Ambient
  • Description:
    Imidocarb dihydrochloride monohydrate

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results