Analytical standard, Ceftiofur hydrochloride, reference material, Honeywell Fluka™
Catalog # HONCR1201-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard, neat
- CAS No.:103980-44-5
- Description:Ceftiofur hydrochloride
- Formula:C₁₉H₁₇N₅O₇S₃ · HCl
- Storage temperature:Ambient
- Description:Ceftiofur hydrochloride
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results