Order Entry
Switzerland
ContactUsLinkComponent
Analytical standard, Inabenfide, reference material, Honeywell Fluka™
Analytical standard, Inabenfide, reference material, Honeywell Fluka™
  HONCR1102-10MG
 :  
Analytical standard, Inabenfide, reference material, Honeywell Fluka™
  HONCR1102-10MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    82211-24-3
  • Description:
    Inabenfide
  • Melting point:
    201…204 °C
  • Formula:
    C₁₉H₁₅ClN₂O₂
  • Storage temperature:
    2…8 °C
  • Description:
    Inabenfide

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results