Order Entry
Switzerland
ContactUsLinkComponent
Analytical standard, Nodularin solution, reference material, Honeywell Fluka™
Analytical standard, Nodularin solution, reference material, Honeywell Fluka™
  HONCR1075-1ML
 :  
Analytical standard, Nodularin solution, reference material, Honeywell Fluka™
  HONCR1075-1ML
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    1 mL
  • Standards:
    Analytical standard, single component solution
  • CAS No.:
    118399-22-7
  • Description:
    Nodularin solution, 10 μg/ml in methanol
  • Melting point:
    –98 °C
  • Formula:
    C₄₁H₆₀N₈O₁₀
  • Storage temperature:
    –20°C
  • Description:
    Nodularin solution
  • Boiling point:
    64…65 °C

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results