Analytical standard, Methiocarb-(N-methyl-d₃), reference material, Honeywell Fluka™
HONCR1039-10MG
:
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:1581694-94-1
- Description:Methiocarb-(N-methyl-d₃)
- Melting point:119 °C
- Formula:C₁₁D₃H₁₂NO₂S
- Storage temperature:Ambient
- Description:Methiocarb-(N-methyl-d₃)
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results