Analytical standard, Clorprenaline, reference material, Honeywell Fluka™
HONCR1023-10MG
:
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:3811-25-4
- Description:Clorprenaline
- Melting point:103…108 °C
- Formula:C₁₁H₁₆ClNO
- Storage temperature:Ambient
- Description:Clorprenaline
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results