(d(CH₂)₅¹,Tyr(Me)²,Thr⁴,Orn⁸,Tyr-NH₂⁹)-Vasotocin Trifluoroacetate salt
BACE4015938.0001
:
- Pk:1 mg
- CAS No.:114056-26-7
- Storage conditions:−20 ±5 °C
- Protein synonyms:OVTA
- Protein/peptide name:(d(CH₂)₅¹,Tyr(Me)²,Thr⁴,Orn⁸,Tyr-NH₂⁹)-Vasotocin Trifluoroacetate salt
- Molecular weight:1154,42
- Molecular formula:C₅₄H₇₉N₁₁O₁₃S₂
The oxytocin antagonist OVTA shows enhanced affinity for rat uterine oxytocin receptors after iodination.
- Sequence: β-Mercapto-β,β-cyclopentamethylene-propionyl-Tyr(Me)-Ile-Thr-Asn-Cys-Pro-Orn-Tyr-NH₂
- Source: synthetic