Order Entry
Switzerland
ContactUsLinkComponent
D-ring anchorage, Protecta®, 3M™
D-ring anchorage, Protecta®, 3M™
Catalog # CPSDAM211
Supplier:  CAPITAL SAFETY
undefined
D-ring anchorage, Protecta®, 3M™
Catalog # CPSDAM211
Supplier:  CAPITAL SAFETY

Specifications

  • Description:
    D-ring anchorage
  • Length:
    50 mm
  • Pk:
    1

Specifications

About this item

Protecta® anchorage D-ring made from 316 stainless steel; 12 mm fixing.

  • Can be used with three load directions
Certifications: EN 795 Class A
Ordering information: Supplied without fixing.