Z-Gln-Gly-OH ≥99% (by TLC)
Supplier: BACHEM BIOCHEMICA
Certificates
About this item
CAS Number:6610-42-0
:
Formula:C₁₅H₁₉N₃O₆
MDL Number:MFCD00055926
:337.332 g/mol
Melting Pt:187 °C
Storage Temperature:Freezer
Specification Test Results
- Identification (TLC):Complies with Authentic Material
- Purity (TLC):99%
- Identification (Elemental Analysis), C:53.4%
- Identification (Elemental Analysis), H:5.7%
- Identification (Elemental Analysis), N:12.5%
- Assay (Elemental Analysis):99.5%
Specification Test Results
Frequently Bought Together