Self-laminating Polyester Cryogenic Laboratory Labels, M211 and M210
Fournisseur: Brady
Certificats
À propos de cet article
Printable high-performance laboratory self-laminating polyester.
- Matt finish
- Ultra-thin construction prevents jamming in racks and centrifuges
Suitable for liquid nitrogen application if material is wrapped around itself.
Frequently Bought Together