Organic reference standard, Bucetin, Supelco®
B4027-10G
: Merck
:
- Pack type:Poly bottle
- Pk:10 g
- Standards:Analytical standard
- CAS No.:1083-57-4
- Description:Bucetin
- Formula:C₁₂H₁₇NO₃
- Description:Bucetin
- Molecular weight (Mw):223,27 g/mol
Analytical standard suitable for HPLC and gas chromatography (GC).