Orders LinkContactUsLinkComponent
Ring light, VisiLED Slim
Ring light, VisiLED Slim
VisiLED slim ring light, FWD 40 - 90 mm and 20 - 40 mm, int.Ø 66 mm, ext.Ø 82 mm, max. 65 klx (at 50 mm FWD) and 45 klx (at 25 mm FWD)
Catalog # SCOC400150
Ring light, VisiLED Slim
Catalog # SCOC400150
Ring light, VisiLED Slim
Ring light, VisiLED Slim
VisiLED slim ring light, FWD 40 - 90 mm and 20 - 40 mm, int.Ø 66 mm, ext.Ø 82 mm, max. 65 klx (at 50 mm FWD) and 45 klx (at 25 mm FWD)
Price
805.00 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents
The VisiLED Slim is a space saving LED ring light, optimal for unconfined object preparation or usage with objective revolvers.
  • Easily adaptable to microscope objectives by means of thumb screw
  • Homogeneous incident or oblique illumination
  • Controllable with all VisiLED controllers (MC750, MC1000, MC1500)
  • Variable working distance ranges possible by exchange of focus optic rings
  • Implementation of brightfield and darkfield applications
  • Segments controllable in different preset modes
Specifications
  • Description:
    VisiLED slim ring light, FWD 40 - 90 mm and 20 - 40 mm, int.Ø 66 mm, ext.Ø 82 mm, max. 65 klx (at 50 mm FWD) and 45 klx (at 25 mm FWD)
  • Seq:
    647