Organic reference standard, chlorbenside, Dr Ehrenstorfer
Catalog # EHERC11150000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pk:0,1 g
- Standards:Reference standard
- CAS No.:103-17-3
- Description:Chlorbenside
- Formula:C₁₃H₁₀Cl₂S
- Description:Chlorbenside
Specifications
About this item
Certifications: Produced in accordance to ISO 17034