Ion exchange chromatography columns, BioPro series
Catalog # YMCAQF00S05-1046WP
Supplier: YMC
Specifications
- Ø int.:4,60 mm
- Particle Size:5 µm
- Description:YMC-BioPro QA-F, non-porous
- Length:100 mm
- Seq:01
- pH range:2 - 12
- Pk:1
Specifications
About this item
YMC-BioPro ion exchange columns are used for the separation of proteins, peptides, and nucleic acids.
- Non-porous particles offer high efficiency and exceptional resolution
- Low operating pressure
YMC-BioPro IEC columns are available in QA and SP chemistries and are based on porous (QA and SP columns) and non-porous (QA-F and SP-F columns) hydrophilic polymer beads with low nonspecific adsorption.