Disinfectants, ClearKlens Tego® 2000 VT25
Supplier: Diversey
Certificates
About this item
TEGO 2000 is a versatile and effective amphoteric-based disinfectant suitable for open plant cleaning applications throughout the food, beverage and dairy industries.
- Ready to use, 0,2 micron filtered, sterile filled and double-bagged
- Validated HPLC and titration test methods available
- Compatible with ClearKlens Cleansinald and ClearKlens Bi-Spore as a rotational process
- Highly effective terminal disinfectant for food industry use
- Effective in soft or hard water
Tego 2000 should be applied after surfaces have been thoroughly cleaned and rinsed. It can be used for disinfecting floors, walls, utensils and other food preparation equipment and is suitable for use in meat and poultry processors, snack foods, dairies, beverage plants and most other types of food processing operations.
Certifications: Passes EN 1276, EN 1650 and EN 13697
Caution: Use biocides safely. Always read the label and product information before use.
Frequently Bought Together