Orders LinkContactUsLinkComponent
General purpose thermometers, enclosed scale
General purpose thermometers, enclosed scale
Supplier:   Amarell
General purpose thermometers, enclosed scale
Supplier:  Amarell
Certificates
General purpose thermometers, enclosed scale
11 Options Available Below
Product Details & Documents
These general purpose standard thermometers are designed for daily use in the laboratory. Capillary prismatic, colourless, total immersion.
Product Family Options
Product Information
  • Range
    Division
    Length
    Filling
    Immersion depth
    Availability
    Price
  • AMARG10000-FL
    General purpose thermometer, capillary lens front, colourless, red special liquid
    General purpose thermometer, capillary lens front, colourless, red special liquid
    −35...+50 °C
    1,0 °C
    260 mm
    Red
    Total
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 30.10
    SFr. 30.10
    1 * 1 ST
       SFr. 30.10
  • AMARG10004-FL
    General purpose thermometer, capillary lens front, colourless, red special liquid
    General purpose thermometer, capillary lens front, colourless, red special liquid
    −10...+60 °C
    1,0 °C
    200 mm
    Red
    Total
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 26.80
    SFr. 26.80
    1 * 1 ST
       SFr. 26.80
  • AMARG10006-FL
    General purpose thermometer, capillary lens front, colourless, red special liquid
    General purpose thermometer, capillary lens front, colourless, red special liquid
    −10...+100 °C
    1,0 °C
    260 mm
    Red
    Total
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 26.80
    SFr. 26.80
    1 * 1 ST
       SFr. 26.80
  • AMARG10008-FL
    General purpose thermometer, capillary lens front, colourless, red special liquid
    General purpose thermometer, capillary lens front, colourless, red special liquid
    −10...+100 °C
    1,0 °C
    260 mm
    Red
    Total
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 31.00
    SFr. 31.00
    1 * 1 ST
       SFr. 31.00
  • AMARG10010-FL
    General purpose thermometer, capillary lens front, colourless, red special liquid
    General purpose thermometer, capillary lens front, colourless, red special liquid
    −10...+150 °C
    1,0 °C
    260 mm
    Red
    Total
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 29.50
    SFr. 29.50
    1 * 1 ST
       SFr. 29.50
  • AMARG10014-FL
    General purpose thermometer, capillary lens front, colourless, red special liquid
    General purpose thermometer, capillary lens front, colourless, red special liquid
    −10...+200 °C
    1,0 °C
    300 mm
    Red
    Total
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 33.60
    SFr. 33.60
    1 * 1 ST
       SFr. 33.60
  • AMARG10016-FL
    General purpose thermometer, capillary lens front, colourless, red special liquid
    General purpose thermometer, capillary lens front, colourless, red special liquid
    −10...+250 °C
    1,0 °C
    300 mm
    Red
    Total
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 49.80
    SFr. 49.80
    1 * 1 ST
       SFr. 49.80
  • AMARG10000-FL-PTFE
    General purpose thermometer, capillary prismatic, colourless, red special liquid, pin-top finish
    General purpose thermometer, capillary prismatic, colourless, red special liquid, pin-top finish
    −35...+50 °C
    1,0 °C
    260 mm
    Red
    Total
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 63.80
    SFr. 63.80
    1 * 1 ST
       SFr. 63.80
  • AMARG10006-FL-PTFE
    General purpose thermometer, capillary prismatic, colourless, red special liquid, pin-top finish
    General purpose thermometer, capillary prismatic, colourless, red special liquid, pin-top finish
    −10...+100 °C
    1,0 °C
    260 mm
    Red
    Total
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 60.50
    SFr. 60.50
    1 * 1 ST
       SFr. 60.50
  • AMARG10010-FL-PTFE
    General purpose thermometer, capillary prismatic, colourless, red special liquid, pin-top finish
    General purpose thermometer, capillary prismatic, colourless, red special liquid, pin-top finish
    −10...+150 °C
    1,0 °C
    260 mm
    Red
    Total
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 63.20
    SFr. 63.20
    1 * 1 ST
       SFr. 63.20
  • AMARG10014-FL-PTFE
    General purpose thermometer, capillary prismatic, colourless, red special liquid, pin-top finish
    General purpose thermometer, capillary prismatic, colourless, red special liquid, pin-top finish
    −10...+200 °C
    1,0 °C
    300 mm
    Red
    Total
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 70.10
    SFr. 70.10
    1 * 1 ST
       SFr. 70.10