Order Entry
Switzerland
ContactUsLinkComponent
R-Phycoerythrin
R-Phycoerythrin
Catalog # OZBIPHY0100
Supplier:  OZ BIOSCIENCES
CAS Number:  
R-Phycoerythrin
Catalog # OZBIPHY0100
Supplier:  OZ BIOSCIENCES
CAS Number:  

Specifications

  • Pk:
    100 µl
  • Protein synonyms:
    R-PE
  • Protein/peptide name:
    R-Phycoerythrin

Specifications

About this item

It is a naturally fluorescent protein from algae and is composed of a 240 kD protein covalently linked to a number of fluorophores called phycobilins. Its exceptional fluorescence characteristics make it a perfect positive control for intracellular protein delivery using OZ Biosciences Pro-DeliverIN reagent. R-phycoerythrin is provided at a 100 μg/ml concentration in DPBS pH 7,4.

  • Absorption maximum: 498/565 nm
  • Emission maximum: 573 nm
  • Extinction coefficient: 1,96×10⁶ M⁻¹ cm⁻¹
  • Quantum yield - 0,84

Store at +4 °C and protect from the light.

Application: Transfection control

Delivery information: Shipped at room temperature.