Bradford protein assay kits (B-Pak)
Supplier: OZ BIOSCIENCES
Certificates
About this item
Bradford Protein Assay Kit (B-Pak) is a straightforward and rapid kit for determining the concentration of proteins in solution. This ready to use kit is provided with 1X reagent and 2 sets of BSA standards which means no dilution, filtration or calculation required. B-Pak is provided with adapted protocols for all experimental conditions – micro and macroassays.
- Accurate determination of protein concentration
- Ready to use prediluted standard protein
- Cheaper than competitors
- Convenient packaging
- Fast and easy to use
Store at +4 °C.
Delivery: Shipped at room temperature.