Magnetic sets
About this item
These magnetic sets have been developped for efficient in vivo transfection or transduction with in vivo Magnetofection technology.
- Can be easily cleaned and decontaminated with 70% ethanol, or by dry or wet sterilisation
- Can be used at room temperature, 37 °C, +4 °C, etc.
- Magnetic properties, distance between magnets and cells and incubation time have been optimized to efficiently concentrate nucleic acids or virus at the targetd site
- Solid, completely reusable, it is a one-time buy
Magnetic properties, distance between magnets and cells and incubation time have been optimized to efficiently concentrate nucleic acids or virus at the targeted site.
3 Options Available Below
Product Details & Documents
These magnetic sets have been developped for efficient in vivo transfection or transduction with in vivo Magnetofection technology.
- Can be easily cleaned and decontaminated with 70% ethanol, or by dry or wet sterilisation
- Can be used at room temperature, 37 °C, +4 °C, etc.
- Magnetic properties, distance between magnets and cells and incubation time have been optimized to efficiently concentrate nucleic acids or virus at the targetd site
- Solid, completely reusable, it is a one-time buy
Magnetic properties, distance between magnets and cells and incubation time have been optimized to efficiently concentrate nucleic acids or virus at the targeted site.
Product Family Options
Product Information
- AvailabilityPrice
- OZBIIV-MAG1Transfection magnets, set of four, Ø 2 mm, 5 mm, 10 mm, and 18×18 mmTransfection magnets, set of four, Ø 2 mm, 5 mm, 10 mm, and 18×18 mmLog in for availability1 * 1 STSFr. 53.61
- OZBIIV-MAG2Square transfection magnets, set of four, 18×18 mm
- OZBIIV-MAG3Cylindrical transfection magnets, sets of four, Ø 2 mm, 5 mm and 10 mmCylindrical transfection magnets, sets of four, Ø 2 mm, 5 mm and 10 mmLog in for availability1 * 1 STSFr. 64.51