Stem transfection reagents, DreamFect™ Stem
Supplier: OZ BIOSCIENCES
Certificates
About this item
DreamFect™ Stem is a powerful transfection reagent allowing multipotent stem cell transfection with high efficiency and very low toxicity. Its specific composition based on TEE-technology allows transfecting embryonic and multipotent stem cells in the presence of serum, maintaining their undifferentiated stage and their capacity to differentiate.
- High transfection efficiency for multipotent stem cells
- Minimised toxicity due to reagent biodegradability and low DNA amount required
- Cell phenotype and differentiation potential are not affected
- Serum compatible
- Simple, ready to use and rapid
Store at –20 °C.
Delivery information: Shipped at room temperature.
Specifications
- Cell specific:Stem cells
- DNA:Yes
- High throughput screening:Yes