siRNA transfection reagent, Lullaby Stem
Supplier: OZ BIOSCIENCES
Certificates
About this item
Lullaby Stem siRNA transfection reagent is ideal for gene silencing in stem cells. Lullaby Stem enables siRNA and miRNA transfection of multipotent and embryonic stem cells with high efficiency and very low toxicity. Its lipid-specific formulation protects small RNA from extracellular degradation, transports them across cell membranes and efficiently releases them into stem cell cytoplasm according to TEE-technology.
- High gene silencing efficiency of multipotent and embryonic stem cells
- Minimised toxicity due to reagent biodegradability and low siRNA/miRNA amount required
- Reliable and reproducible gene knockdown results
- Serum and antibiotics compatible
Store at +4 °C.
Delivery information: Shipped at room temperature.
Specifications
- Cell specific:Stem cells
- shRNA:Yes
- siRNA:Yes