HPTLC plates, nano silica gel 60, unmodified layers, Nano-SIL HD
Anbieter: MACHEREY-NAGEL
Zertifikate
Über diesen Artikel
Unmodified HPTLC silica layers with "High Definition" properties for HPTLC.
- Nano silica gel (particle size 2 to 10 μm)
- Optimised binder system
- High luminosity and brilliant staining properties
Besides a good wettability for precise colorisation results the Nano-SIL HD glass plates allows sharper separations, shorter developing times and shorter migration distances compared to standard silica layers. High suitability for trace analyses resulting from a UV indicator with increased brilliance and a low-noise background of the layer.
Nano silica 60: Specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 2 – 10 μm.
Frequently Bought Together