Order Entry
Switzerland
ContactUsLinkComponent
Yeast extract, powder, ultrapure
Yeast extract, powder, ultrapure
Catalog # J23547.A1
CAS Number:  
Yeast extract, powder, ultrapure
Catalog # J23547.A1
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Description:
    Yeast Extract, Powder
  • Storage conditions:
    Ambient

Specifications

About this item

  • Assay Protein (TN×6.25): 57 - 72%
  • Assay Salmonella: Negative
  • Assay Standard Plate Count: ≤15000/g
  • Residual Water(at Time of Packaging): ≤6%