Protein Prep syringe filters for ӒKTA™ systems, Whatman™
Supplier: Whatman products (Cytiva)
Certificates
About this item
Protein prep syringe filters for ÄKTA™ protein purification systems are designed for optimal preparation of protein samples prior to chromatography.
- Hydrophilic membrane made of regenerated cellulose exhibits low protein binding for sample preservation
- Lot-to-lot certified to have low levels of extractables that could otherwise interfere with work
- Filtration of samples prior to runs demonstrated to reduce column degrading particulate matter
High yield protein purification and accurate protein analysis both depend on a quality sample being introduced to the chromatography column. The protein prep syringe filter has been designed specifically to optimise these outcomes.
Protein prep syringe filters are ready-to-use with polycarbonate housing and a regenerated cellulose membrane that is low protein binding and broadly compatible with common solvents. Syringe filtration has been shown to reduce debris residue in the column that could otherwise impact performance and column life.
Frequently Bought Together