SPE columns, hydrocarbons analysis, CHROMABOND® Na₂SO₄/Florisil®
Supplier: MACHEREY-NAGEL
Certificates
About this item
CHROMABOND® Na₂SO₄/Florisil® is a combination phase for SPE of hydrocarbons from water.
- Sodium sulfate phase for removal of water traces from cyclohexane extract
- Florisil® phase for enrichment of hydrocarbons
Application: Extraction of hydrocarbons from water acc. to ISO DIS 9377-4 / DIN H-53.
Specifications
- Catalog No:
- MANA730249G
- MANA730249G.250
- MANA730249
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
Specifications
Frequently Bought Together