Karl Fischer reagents, buffer substances, HYDRANAL™, Fluka™
Anbieter: Honeywell Chemicals
Zertifikate
Zertifikate
Über diesen Artikel
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Anbieter: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Anbieter: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
Glass microfibre filters without binder, grade GF/C RTU, Whatman™
Anbieter: Whatman products (Cytiva)
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Anbieter: VWR Chemicals
Acetone ≥99%, TECHNICAL
Anbieter: VWR Chemicals
Single use syringes, 2-piece, Injekt® Solo
Anbieter: B. Braun
Laboratory trays, coloured
Anbieter: MULTIROIR
VWR®, Nitrile Gloves
Anbieter: VWR Collection
pH indicator strips, MQuant®, Supelco®
Anbieter: Merck
Potassium dihydrogen phosphate 99+%, extra pure
Anbieter: Thermo Fisher Scientific