Order Entry
Switzerland
ContactUsLinkComponent
Cellulose MN 2100ff adsorbents for column chromatography (cellulose MN 2100 defatted)
Cellulose MN 2100ff adsorbents for column chromatography (cellulose MN 2100 defatted)
Catalog # MANA815070.1
Supplier:  MACHEREY-NAGEL
CAS Number:  
Cellulose MN 2100ff adsorbents for column chromatography (cellulose MN 2100 defatted)
Catalog # MANA815070.1
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :342.3 g/mol
CAS Number:9004-34-6
Formula:(C₁₂H₂₂O₁₁)n
MDL Number:MFCD00081512