TLC education advanced kit, micro-set F3
Anbieter: MACHEREY-NAGEL
Zertifikate
Über diesen Artikel
Advanced kit for science education.
- All chemicals included for the separation of:
- Analgetics (pain relievers)
- Drugs as shown for cinchona bark
Together with the materials from TLC micro-Set M Aspirin® and Thomapyrin® tablets as well as the drug cinchona bark can be separated and visualised by spray reagents (e.g., reagent according to Dragendorff-Munier).
Caution: * These products contain harmful substances which must be specially labelled as hazardous.
Frequently Bought Together