Order Entry
Switzerland
ContactUsLinkComponent
 
 
Sodium hydroxide ≥98.0% (by acidimetry), pellets ACS, ISO for analysis
 
 
Sodium hydroxide ≥98.0% (by acidimetry), pellets ACS, ISO for analysis
 

 

  • Minimum Assay (Acidim.):
    98.0%
  • Insoluble Matter in H₂O:
    Max. 0.005%
  • Chloride (Cl):
    Max. 0.001%
  • Nitrogen Compounds (as N):
    Max. 0.0005%
  • Phosphate (PO₄):
    Max. 0.0005%
  • Sulphate (SO₄):
    Max. 0.002%
  • Silicate SiO₂:
    Max. 0.001%
  • Carbonate (as Na₂CO₃):
    Max. 1.0%
  • Heavy Metals (as Pb):
    Max. 0.0005%
  • Al:
    Max. 0.0005%
  • As:
    Max. 0.00004%
  • Ca:
    Max. 0.0005%
  • Cd:
    Max. 0.0005%
  • Co:
    Max. 0.0005%
  • Cu:
    Max. 0.0005%
  • Fe:
    Max. 0.0005%
  • Hg:
    Max. 0.00001%
  • K:
    Max. 0.02%
  • MG:
    Max. 0.0005%
  • Mn:
    Max. 0.0005%
  • Ni:
    Max. 0.0005%
  • PB:
    Max. 0.0005%
  • Zn:
    Max. 0.001%

 

Frequently Bought Together