Order Entry
Switzerland
ContactUsLinkComponent
Karl fischer reagents, Fluka™
Karl fischer reagents, Fluka™
Certificates
undefined
Karl fischer reagents, Fluka™
Certificates

Specifications

  • Flash point:
    16 °C
  • Boiling point:
    >60 °C

Specifications

Related Information

Frequently Bought Together