ContactUsLinkComponent
Immersion oil, BioChemica
Catalog # A3494.0500
Anbieter:  PanReac AppliChem
CAS-Nummer:  
Immersion oil, BioChemica
Catalog # A3494.0500
Anbieter:  PanReac AppliChem
CAS-Nummer:  
Technische Daten
  • Pk:
    500 mL
  • Description:
    Immersion oil
Technische Daten
Über diesen Artikel
Biochemical grade immersion oil for microscopy.
  • Free of PCBs
  • Stored at room temperature