Order Entry
Switzerland
ContactUsLinkComponent
Benchtop biohazard disposal can with motion sensor lid, SP Bel-Art
Benchtop biohazard disposal can with motion sensor lid, SP Bel-Art
Catalog # 212-0732
undefined
Benchtop biohazard disposal can with motion sensor lid, SP Bel-Art
Catalog # 212-0732

Specifications

  • Ø×H:
    124×170 mm
  • Capacity:
    1,5 l
  • Description:
    Biohazard disposal can
  • Pk:
    1

Specifications

About this item

Compact benchtop disposal can contains odours and is perfect for small items such as pipette tips and microcentrifuge tubes.

  • Automatic opening lid provides hands free use; closes automatically and securely for a more sanitary environment
  • Holds a 25×25 cm autoclavable biohazard disposal bag
  • Includes a polypropylene liner with handle for easy emptying of contents
  • Aesthetic and durable; stainless steel body and chrome lid are resistant to bleach and easy to clean
Ordering information: Requires (3) AAA batteries (not included).