Order Entry
Switzerland
ContactUsLinkComponent
Acetonitrile ≥99.8% (GC, on anhydrous basis), PESTINORM® Supra Trace for organic trace analysis
Flammable
Irritant
Acetonitrile ≥99.8% (GC, on anhydrous basis), PESTINORM® Supra Trace for organic trace analysis
Catalog # 85935.320
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  
undefined
Flammable
Irritant
Acetonitrile ≥99.8% (GC, on anhydrous basis), PESTINORM® Supra Trace for organic trace analysis
Catalog # 85935.320
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Appearance:
    Clear, colourless liquid
  • Assay (GC, on anhydrous basis):
    min. 99.8 %
  • Identity (IR):
    Conforms
  • Residue after evaporation:
    max. 0.0003 % w/w
  • Water (KF):
    max. 0.05 % w/w
  • Color (APHA, Pt-Co):
    max. 10
  • GC/ECD Dioxins, Furans & PCB's:
    max. 3 pg/ml
  • GC/ECD any Pesticide (as Lindane):
    max. 3 pg/ml
  • GC/ECD 1,2,4-TCB to daca-PCB (asLindane):
    max. 3 pg/ml
  • GC/FID C10 to C40 (as n-tetradecane):
    max. 3 ng/ml
  • GC/MSD C10 to C40 (as Decane; 30-600amu):
    max. 3 ng/ml

Specification Test Results

About this item

 :
 :41.05 g/mol
Boiling Pt:81.6 °C (1013 hPa)
Melting Pt:–45.7 °C
ADR:3,II
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
EINECS:200-835-2
UN:1648
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878