ESD safety ankle boots, lace-up, Biomex Dynamics, Till Boa
: ELTEN
ESD safety ankle boots made from water resistant cowhide leather upper, a full padded bellows tongue an extremely flexible, ergonomic, S-shaped, dual-density TPU/PU sole (Biomex Dynamics).
- Full length fleece inlay sole (ESD Pro), black
- Replaceable insole; orthopaedic insoles possible
- Soft fleece ESD insole
- Anatomically-shaped insole and ankle support for improved stability
- Lace- and buckle-free Boa® system can be opened and closed with just one hand
- TPU scuff cap protects the upper
- Puncture resistant steel midsole
- Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC, HI, HRO.
- Catalog No:
- ELTN76651/43
- ELTN76651/44
- ELTN76651/40
- ELTN76651/41
- ELTN76651/38
- ELTN276651/49
- ELTN76651/39
- ELTN76651/47
- ELTN176651/36
- ELTN76651/37
- ELTN76651/48
- ELTN76651/45
- ELTN76651/46
- ELTN76651/42
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
Frequently Bought Together