ESD safety shoes, lace-up, Biomex Dynamics, Impulse Green
Supplier: ELTEN
Certificates
About this item
Metal- and leather-free, lightweight ESD safety shoe made from perforated microfibre and reflective materials, skin friendly textile lining, a full padded bellows tongue and collar plus an extremely flexible, egronomic S-shaped, dual-density TPU/PU sole (Biomex Dynamics).
- Climate-regulating, breathable textile lining
- Anatomically-shaped insole supports the natural heel-to-toe foot movement
- Replaceable insole; orthopaedic insoles possible
- Full length ESD inlay sole
- Abrasion-proof scuff cap
- Protective composite toecap (200 J)
- Metal-free, puncture resistant midsole
Ideal for use in dry work areas such as industry, storage, transport, or assembly and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/green
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC.
Specifications
- Catalog No:
- ELTN722551/37
- ELTN722551/36
- ELTN722551/38
- ELTN722551/40
- ELTN722551/39
- ELTN722551/42
- ELTN722551/44
- ELTN722551/41
- ELTN722551/46
- ELTN722551/43
- ELTN722551/45
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001