Western-Re-Probe™ reagent, Calbiochem®
Catalog # WB59-100
Supplier: Merck Millipore (Oncogene)
CAS Number:
Specifications
- Pk:100 mL
- Description:Western-Re-Probe™ reagent
Specifications
About this item
This Western-Re-Probe™ reagent is validated for use in immunoblotting.
- Immunoblotting analysed repeatedly with multiple probes
- Saving on hard to get reagents and samples
- Correction and reanalysis of unsatisfactory Western blots
- Saving time spent in gel rerunning and transfer processes
Designed to dissociate and remove antibodies from membrane-bound proteins without destroying antigenic binding capacity. Recommended for stripping chemiluminescent or radioisotopic signals from blots. Not recommended for stripping colour-producing Eestern blots.
Supplied as a 5X solution sufficient for stripping 25 to 30 standard blots.
Delivery information: Supplied as a 5X solution sufficient for stripping 25 to 30 standard blots.
Caution: Western-Re-Probe™ reagent can be used for stripping and re-probing membranes 2-3 times. Some loss of signal may be noticed after second cycle of stripping and re-probing.