Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Standard redox solution, SensION+

Standard redox solution, SensION+

SensION+, standard redox solution, 220 mV, 250 mL
Catalog # HACHLZW9400.99
Supplier:   Hach
Standard redox solution, SensION+
Catalog # HACHLZW9400.99
Supplier:  Hach
CAS Number:  
Standard redox solution, SensION+
Standard redox solution, SensION+

SensION+, standard redox solution, 220 mV, 250 mL

CAS Number:  
Price
SFr. 66.40
Log in for availability
1 * 250 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    SensION+, standard redox solution, 220 mV

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.