ContactUsLinkComponent
Aprons, zetDress®
Aprons, zetDress®
Catalog # ZVGA80160
Aprons, zetDress®
Catalog # ZVGA80160
Specifications
  • Anti-static:
    +
  • Garment application:
    General Purpose,
    Laboratory,
    Industrial,
    Food
  • Colour:
    White
  • Pk:
    100
  • W×L:
    750×1250 mm
Specifications
About this item
Aprons, made from PE foil, are suitable for sensitive work areas, such an the food processing industry, the sustainable maintenance of a hygienic environment, is indispensable.
  • Robust and disposable
  • Hammered foil
  • Individually folded for easy removal
Protects from biological contaminants, radioactive particles and liquid chemicals, also anti-static, spray-proof and particle-tight.